COMH4211AF28 Combi-Blocks, Inc.

Methyl 2,4,6-trifluoro-3-nitrobenzoate

  • MDL number MFCD31579482
  • Cas Number : 125568-72-1
  • Molecular Weight : 235.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2768

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2768
Availability details for
Quantity
  • Availability
  • Add to Cart