COMH4211AF2A Combi-Blocks, Inc.

(3-Chloro-2-propoxy-benzyl)-methyl-amine

  • MDL number MFCD24611653
  • Cas Number : 894851-33-3
  • Molecular Weight : 213.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2770

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2770
Availability details for
Quantity
  • Availability
  • Add to Cart