COMH4211AF2D Combi-Blocks, Inc.

6-(4-Iodo-phenoxy)-nicotinamide

  • MDL number MFCD13230460
  • Cas Number : 676495-46-8
  • Molecular Weight : 340.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 6-(4-iodophenoxy)nicotinamide
Inchi Code 1S/C12H9IN2O2/c13-9-2-4-10(5-3-9)17-11-6-1-8(7-15-11)12(14)16/h1-7H,(H2,14,16)
Inchi Key NKKPJEGINOPHNB-UHFFFAOYSA-N

Description

Purity 95%
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2787

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2787
Availability details for
Quantity
  • Availability
  • Add to Cart