COMH4211AF2E Combi-Blocks, Inc.

4-[2-[(6-Bromo-3-pyridyl)oxy]ethyl]morpholine

  • MDL number MFCD31613888
  • Cas Number : 1660157-38-9
  • Molecular Weight : 287.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2788

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2788
Availability details for
Quantity
  • Availability
  • Add to Cart