COMH4211AF30 Combi-Blocks, Inc.

4-(4-Hydroxy-phenoxy)-piperidine-1-carboxylic acid tert-butyl ester

  • MDL number MFCD23110521
  • Cas Number : 681484-29-7
  • Molecular Weight : 293.36

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name tert-butyl 4-(4-hydroxyphenoxy)piperidine-1-carboxylate
Inchi Code 1S/C16H23NO4/c1-16(2,3)21-15(19)17-10-8-14(9-11-17)20-13-6-4-12(18)5-7-13/h4-7,14,18H,8-11H2,1-3H3
Inchi Key WKRWQQJCOQHQBH-UHFFFAOYSA-N

Description

Purity 95%
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2793

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2793
Availability details for
Quantity
  • Availability
  • Add to Cart