COMH4211AF31 Combi-Blocks, Inc.

3-Methoxy-4-(4-methoxy-benzyloxy)-benzylamine

  • MDL number MFCD30803476
  • Cas Number : 1183446-64-1
  • Molecular Weight : 273.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2794

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2794
Availability details for
Quantity
  • Availability
  • Add to Cart