COMH4211AF32 Combi-Blocks, Inc.

4-[(Cyclohexanesulfonyl)methyl]aniline

  • MDL number MFCD13161241
  • Cas Number : 1184585-00-9
  • Molecular Weight : 253.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2796

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2796
Availability details for
Quantity
  • Availability
  • Add to Cart