COMH4211AF34 Combi-Blocks, Inc.

2-Amino-5-cyclopropyl-benzoic acid methyl ester

  • MDL number MFCD24642124
  • Cas Number : 1119088-76-4
  • Molecular Weight : 191.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2800

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2800
Availability details for
Quantity
  • Availability
  • Add to Cart