COMH4211AF35 Combi-Blocks, Inc.

[4-(4-Carbamoyl-3-fluoro-phenyl)-piperazin-1-yl]-acetic acid benzyl ester

  • MDL number MFCD31583320
  • Cas Number : 2270907-84-9
  • Molecular Weight : 371.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2807

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2807
Availability details for
Quantity
  • Availability
  • Add to Cart