COMH4211AF36 Combi-Blocks, Inc.

2-Fluoro-4-(2,2,2-trifluoroethoxy)benzonitrile

  • MDL number MFCD23903693
  • Cas Number : 1499400-23-5
  • Molecular Weight : 219.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2808

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2808
Availability details for
Quantity
  • Availability
  • Add to Cart