COMH4211AF37 Combi-Blocks, Inc.

1-Bromo-3-[(2-chloroethyl)sulfanyl]benzene

  • MDL number MFCD17225283
  • Cas Number : 1341412-54-1
  • Molecular Weight : 251.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2809

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2809
Availability details for
Quantity
  • Availability
  • Add to Cart