COMH4211AF39 Combi-Blocks, Inc.

3-Fluoro-5-methoxy-4-nitrobenzoic acid

  • MDL number MFCD20527807
  • Cas Number : 1137869-93-2
  • Molecular Weight : 215.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2812

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2812
Availability details for
Quantity
  • Availability
  • Add to Cart