COMH4211AF3B Combi-Blocks, Inc.

1-(2-tert-Butylphenyl)piperazine dihydrochloride

  • MDL number MFCD31613725
  • Cas Number : 1252657-79-6
  • Molecular Weight : 291.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1-(2-(tert-butyl)phenyl)piperazine dihydrochloride
Inchi Code 1S/C14H22N2.2ClH/c1-14(2,3)12-6-4-5-7-13(12)16-10-8-15-9-11-16;;/h4-7,15H,8-11H2,1-3H3;2*1H
Inchi Key UMSTWQSYHCXZHI-UHFFFAOYSA-N

Description

Purity 95%
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2815

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2815
Availability details for
Quantity
  • Availability
  • Add to Cart