COMH4211AF3C Combi-Blocks, Inc.

(2-Cyano-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)cyclopropanecarboxamide

  • MDL number MFCD28899678
  • Cas Number : 1403248-60-1
  • Molecular Weight : 312.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2817

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2817
Availability details for
Quantity
  • Availability
  • Add to Cart