COMH4211AF3D Combi-Blocks, Inc.

3-Fluoro-4-(2-methoxyethoxy)benzaldehyde

  • MDL number MFCD11156477
  • Cas Number : 942935-47-9
  • Molecular Weight : 198.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2818

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2818
Availability details for
Quantity
  • Availability
  • Add to Cart