COMH4211AF3E Combi-Blocks, Inc.

Trans 4-tert-butoxycarbonylamino-cyclohexanecarboxylic acid 2,5-dioxo-pyrrolidin-1-yl ester

  • MDL number MFCD30377249
  • Cas Number : 2365390-61-8
  • Molecular Weight : 340.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2820

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2820
Availability details for
Quantity
  • Availability
  • Add to Cart