COMH4211AF40 Combi-Blocks, Inc.

4-(4-Amino-phenylsulfanylmethyl)-piperidine-1-carboxylic acid tert-butyl ester

  • MDL number MFCD25973703
  • Cas Number : 1432062-24-2
  • Molecular Weight : 322.5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2830

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2830
Availability details for
Quantity
  • Availability
  • Add to Cart