COMH4211AF42 Combi-Blocks, Inc.

5-Fluoro-2-morpholinonicotinohydrazide

  • MDL number MFCD31613719
  • Cas Number : 2070855-98-8
  • Molecular Weight : 240.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2835

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2835
Availability details for
Quantity
  • Availability
  • Add to Cart