COMH4211AF43 Combi-Blocks, Inc.

4-Fluoro-5-methoxy-2-methyl-benzoic acid

  • MDL number MFCD23712114
  • Cas Number : 1427382-28-2
  • Molecular Weight : 184.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2836

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2836
Availability details for
Quantity
  • Availability
  • Add to Cart