COMH4211AF44 Combi-Blocks, Inc.

5-Bromo-2-isopropoxy-n-methyl-benzamide

  • MDL number MFCD31628785
  • Cas Number : 2270910-95-5
  • Molecular Weight : 272.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2837

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2837
Availability details for
Quantity
  • Availability
  • Add to Cart