COMH4211AF45 Combi-Blocks, Inc.

(S)-tert-Butyl 4-(4-amino-2-fluorophenyl)-3-methylpiperazine-1-carboxylate

  • MDL number MFCD31579451
  • Cas Number : 1679381-26-0
  • Molecular Weight : 309.38

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name tert-butyl (S)-4-(4-amino-2-fluorophenyl)-3-methylpiperazine-1-carboxylate
Inchi Code 1S/C16H24FN3O2/c1-11-10-19(15(21)22-16(2,3)4)7-8-20(11)14-6-5-12(18)9-13(14)17/h5-6,9,11H,7-8,10,18H2,1-4H3/t11-/m0/s1
Inchi Key JLULNKVXYANZET-NSHDSACASA-N

Description

Purity 95%
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2838

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2838
Availability details for
Quantity
  • Availability
  • Add to Cart