COMH4211AF46 Combi-Blocks, Inc.

2-Iodo-3-(propan-2-yloxy)pyridine

  • MDL number MFCD28670141
  • Cas Number : 1296206-79-5
  • Molecular Weight : 263.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2839

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2839
Availability details for
Quantity
  • Availability
  • Add to Cart