COMH4211AF47 Combi-Blocks, Inc.

4-Dimethylaminomethyl-bicyclo[2.2.2]octane-1-carboxylic acid hydrochloride

  • MDL number MFCD31381612
  • Cas Number : 2203140-31-0
  • Molecular Weight : 247.8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2840

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2840
Availability details for
Quantity
  • Availability
  • Add to Cart