COMH4211AF49 Combi-Blocks, Inc.

4-(4-Methoxy-benzyloxy)-1h-pyrazole

  • MDL number MFCD31579523
  • Cas Number : 1893853-00-3
  • Molecular Weight : 204.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2842

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2842
Availability details for
Quantity
  • Availability
  • Add to Cart