COMH4211AF4A Combi-Blocks, Inc.

3-Iodo-2-pyrrolidin-1-ylpyridine

  • MDL number MFCD13195614
  • Cas Number : 1235440-37-5
  • Molecular Weight : 274.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2843

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2843
Availability details for
Quantity
  • Availability
  • Add to Cart