COMH4211AF4B Combi-Blocks, Inc.

(3-Chloro-4-methyl-phenoxy)-acetic acid

  • MDL number MFCD01312573
  • Cas Number : 588-21-6
  • Molecular Weight : 200.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2844

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2844
Availability details for
Quantity
  • Availability
  • Add to Cart