COMH4211AF4C Combi-Blocks, Inc.

1-[5-Bromo-2-(2,2,2-trifluoro-ethoxy)-phenyl]-ethylamine

  • MDL number MFCD16753349
  • Cas Number : 1250558-42-9
  • Molecular Weight : 298.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1-[5-bromo-2-(2,2,2-trifluoroethoxy)phenyl]ethanamine
Inchi Code 1S/C10H11BrF3NO/c1-6(15)8-4-7(11)2-3-9(8)16-5-10(12,13)14/h2-4,6H,5,15H2,1H3
Inchi Key JDXZEQLNFOYULO-UHFFFAOYSA-N

Description

Purity 95%
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2846

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2846
Availability details for
Quantity
  • Availability
  • Add to Cart