COMH4211AF4F Combi-Blocks, Inc.

Beta-alanine, n-(3-phenoxybenzoyl)-

  • MDL number MFCD31381715
  • Cas Number : 133639-10-8
  • Molecular Weight : 285.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2852

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2852
Availability details for
Quantity
  • Availability
  • Add to Cart