COMH4211AF50 Combi-Blocks, Inc.

Benzyl 6-(hydrazinecarbonyl)pyridin-2-ylcarbamate

  • MDL number MFCD26393028
  • Cas Number : 1262043-89-9
  • Molecular Weight : 286.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2854

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2854
Availability details for
Quantity
  • Availability
  • Add to Cart