COMH4211AF51 Combi-Blocks, Inc.

N-(3-Amino-5-(trifluoromethyl)phenyl)methanesulfonamide

  • MDL number MFCD26393308
  • Cas Number : 873056-09-8
  • Molecular Weight : 254.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2856

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2856
Availability details for
Quantity
  • Availability
  • Add to Cart