COMH4211AF53 Combi-Blocks, Inc.

3-(4-Bromo-2-fluoro-phenoxymethyl)-benzoic acid

  • MDL number MFCD12631946
  • Cas Number : 1040077-27-7
  • Molecular Weight : 325.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2858

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2858
Availability details for
Quantity
  • Availability
  • Add to Cart