COMH4211AF54 Combi-Blocks, Inc.

1-Butylcyclopropane-1-sulfonamide

  • MDL number MFCD28959167
  • Cas Number : 681808-60-6
  • Molecular Weight : 177.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2861

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2861
Availability details for
Quantity
  • Availability
  • Add to Cart