COMH4211AF56 Combi-Blocks, Inc.

N-Cyclopropyl-4-hydroxy-2-methylbenzamide

  • MDL number MFCD29954322
  • Cas Number : 1695438-98-2
  • Molecular Weight : 191.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2864

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2864
Availability details for
Quantity
  • Availability
  • Add to Cart