COMH4211AF58 Combi-Blocks, Inc.

2-(3-Trityloxypropyloxy)-ethanol

  • MDL number MFCD31618937
  • Cas Number : 1020728-52-2
  • Molecular Weight : 362.5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2867

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2867
Availability details for
Quantity
  • Availability
  • Add to Cart