COMH4211AF5A Combi-Blocks, Inc.

6-Amino-n-ethylpyridine-3-carboxamide

  • MDL number MFCD16678973
  • Cas Number : 1248024-71-6
  • Molecular Weight : 165.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2869

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2869
Availability details for
Quantity
  • Availability
  • Add to Cart