COMH4211AF5B Combi-Blocks, Inc.

(3-Ethoxy-5-hydroxymethyl-phenyl)-methanol

  • MDL number MFCD31628809
  • Cas Number : 277324-97-7
  • Molecular Weight : 182.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2872

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2872
Availability details for
Quantity
  • Availability
  • Add to Cart