COMH4211AF5C Combi-Blocks, Inc.

1-[4-(2-Methylpropoxy)phenyl]propan-2-one

  • MDL number MFCD19249660
  • Cas Number : 1266949-83-0
  • Molecular Weight : 206.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2873

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2873
Availability details for
Quantity
  • Availability
  • Add to Cart