COMH4211AF5D Combi-Blocks, Inc.

Ethanone,1-[4-(2-hydroxyethyl)-1-piperidinyl]-

  • MDL number MFCD11617079
  • Cas Number : 15871-63-3
  • Molecular Weight : 171.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2878

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2878
Availability details for
Quantity
  • Availability
  • Add to Cart