COMH4211AF5E Combi-Blocks, Inc.

(E)-Ethyl 4-aminobut-2-enoate hydrochloride

  • MDL number MFCD28385438
  • Cas Number : 2365421-25-4
  • Molecular Weight : 165.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2884

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2884
Availability details for
Quantity
  • Availability
  • Add to Cart