COMH4211AF63 Combi-Blocks, Inc.

(4-Bromo-2-fluorophenyl)(4-methylpiperazin-1-yl)methanone

  • MDL number MFCD09942268
  • Cas Number : 924642-60-4
  • Molecular Weight : 301.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2895

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2895
Availability details for
Quantity
  • Availability
  • Add to Cart