COMH4211AF65 Combi-Blocks, Inc.

2-(3-(4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)propyl)isoindoline-1,3-dione

  • MDL number MFCD31561556
  • Cas Number : 1628595-38-9
  • Molecular Weight : 391.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2897

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2897
Availability details for
Quantity
  • Availability
  • Add to Cart