COMH4211AF66 Combi-Blocks, Inc.

((2R,3R,4S,5R)-6-Allyloxy-3,4,5-tris-benzyloxy-tetrahydro-pyran-2-yl)-methanol

  • MDL number MFCD31618884
  • Cas Number : 54606-68-7
  • Molecular Weight : 490.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2901

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2901
Availability details for
Quantity
  • Availability
  • Add to Cart