COMH4211AF67 Combi-Blocks, Inc.

1-(6-Bromopyridin-2-yl)-4-(2,2,2-trifluoroethyl)piperazine

  • MDL number MFCD19554855
  • Cas Number : 1486426-73-6
  • Molecular Weight : 324.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2911

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2911
Availability details for
Quantity
  • Availability
  • Add to Cart