COMH4211AF68 Combi-Blocks, Inc.

5-Bromo-2-propoxyaniline

  • MDL number MFCD08687066
  • Cas Number : 946786-50-1
  • Molecular Weight : 230.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2914

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2914
Availability details for
Quantity
  • Availability
  • Add to Cart