COMH4211AF69 Combi-Blocks, Inc.

3-((4-Bromo-3,5-dimethylphenoxy)methyl)-3-methyloxetane

  • MDL number MFCD31583095
  • Cas Number : 1458652-56-6
  • Molecular Weight : 285.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2915

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2915
Availability details for
Quantity
  • Availability
  • Add to Cart