COMH4211AF6A Combi-Blocks, Inc.

Cis trans (4-nitro-cyclohexyl)-carbamic acid tert-butyl ester

  • MDL number MFCD31614088
  • Cas Number : 2270907-11-2
  • Molecular Weight : 244.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2916

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2916
Availability details for
Quantity
  • Availability
  • Add to Cart