COMH4211AF6B Combi-Blocks, Inc.

1-(Dimethylamino)cyclopentane-1-carboxylic acid

  • MDL number MFCD12064743
  • Cas Number : 933690-12-1
  • Molecular Weight : 157.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2919

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2919
Availability details for
Quantity
  • Availability
  • Add to Cart