COMH4211AF6C Combi-Blocks, Inc.

1,1-Dimethylethyl 5-bromo-4-chloro-2,3-dihydro-1h-indole-1-carboxylate

  • MDL number MFCD31583093
  • Cas Number : 1337533-81-9
  • Molecular Weight : 332.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2920

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2920
Availability details for
Quantity
  • Availability
  • Add to Cart