COMH4211AF6D Combi-Blocks, Inc.

5-tert-Butoxycarbonylamino-4-fluoro-2-methyl-benzoic acid

  • MDL number MFCD31579288
  • Cas Number : 658085-43-9
  • Molecular Weight : 269.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2921

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2921
Availability details for
Quantity
  • Availability
  • Add to Cart