COMH4211AF6E Combi-Blocks, Inc.

1-[4-(4-Chloro-3-trifluoromethyl-phenoxy)-phenyl]-ethylamine

  • MDL number MFCD31614126
  • Cas Number : 2270907-04-3
  • Molecular Weight : 315.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2924

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2924
Availability details for
Quantity
  • Availability
  • Add to Cart